gRNA Design Details
- Target DNA Sequence
- AGGCGCCCGAATACAGGTTC
- PAM Sequence
- AGG
- Target locus
-
Chr.4: 148602729 - 148602751 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
91.55/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Masp2
- Entrez Gene ID
-
17175
- Gene Name
- mannan-binding lectin serine peptidase 2
- Gene Aliases
-
MASP-2, MAp19
- Chromosome Location
-
Chr.4: 148594877 - 148615482 on GRCm38
- UniGene ID
- Mm.378962
- Species
-
Mouse