gRNA Design Details
- Target DNA Sequence
- GAGTTTGGTGGCCACACTTC
- PAM Sequence
- TGG
- Target locus
-
Chr.4: 148602695 - 148602717 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
85.88/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Masp2
- Entrez Gene ID
-
17175
- Gene Name
- mannan-binding lectin serine peptidase 2
- Gene Aliases
-
MASP-2, MAp19
- Chromosome Location
-
Chr.4: 148594877 - 148615482 on GRCm38
- UniGene ID
- Mm.378962
- Species
-
Mouse