gRNA Design Details
- Target DNA Sequence
- AGACATCAGCTCATCTACGC
- PAM Sequence
- TGG
- Target locus
-
Chr.8: 54157001 - 54157023 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
83.7/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Vegfc
- Entrez Gene ID
-
22341
- Gene Name
- vascular endothelial growth factor C
- Gene Aliases
-
AW228853, VEGF-C
- Chromosome Location
-
Chr.8: 54077532 - 54186454 on GRCm38
- UniGene ID
- Mm.1402
- Species
-
Mouse