gRNA Design Details

Target DNA Sequence
TTCTCGCTCCGTAGTAGCCG
PAM Sequence
TGG
Target locus
Chr.17: 46031689 - 46031711 on GRCm38
Strand
Reverse
Application
Gene Knock-Out
Score: 92.91/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Vegfa
Entrez Gene ID
22339
Gene Name
vascular endothelial growth factor A
Gene Aliases
Vegf, Vpf
Chromosome Location
Chr.17: 46016993 - 46032377 on GRCm38
UniGene ID
Mm.282184
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001025250.3 NP_001020421.2 1
NM_001025257.3 NP_001020428.2 1
NM_001287056.1 NP_001273985.1 1
NM_001287057.1 NP_001273986.1 1
NM_001287058.1 NP_001273987.1 1
NM_009505.4 NP_033531.3 1