gRNA Design Details
- Target DNA Sequence
- GTCACCTCTACAATACCGTC
- PAM Sequence
- CGG
- Target locus
-
Chr.11: 53433728 - 53433750 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
92.64/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gdf9
- Entrez Gene ID
-
14566
- Gene Name
- growth differentiation factor 9
- Gene Aliases
-
Gdf-9
- Chromosome Location
-
Chr.11: 53430878 - 53437903 on GRCm38
- UniGene ID
- Mm.490461
- Species
-
Mouse