gRNA Design Details
- Target DNA Sequence
- CGTATTCACAGAGATCGTGC
- PAM Sequence
- TGG
- Target locus
-
Chr.19: 45737235 - 45737257 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.13/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fgf8
- Entrez Gene ID
-
14179
- Gene Name
- fibroblast growth factor 8
- Gene Aliases
-
Aigf, Fgf-8
- Chromosome Location
-
Chr.19: 45736798 - 45742935 on GRCm38
- UniGene ID
- Mm.4012
- Species
-
Mouse