gRNA Design Details
- Target DNA Sequence
- CCAGACCGAACGTGAAGACG
- PAM Sequence
- AGG
- Target locus
-
Chr.9: 120052193 - 120052215 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
92.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Cx3cr1
- Entrez Gene ID
-
13051
- Gene Name
- chemokine (C-X3-C motif) receptor 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 120048683 - 120068296 on GRCm38
- UniGene ID
- Mm.44065
- Species
-
Mouse