gRNA Design Details
- Target DNA Sequence
- CTTTAGTACATGAAAGCAGC
- PAM Sequence
- AGG
- Target locus
-
Chr.11: 82116054 - 82116076 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
83.68/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ccl8
- Entrez Gene ID
-
20307
- Gene Name
- chemokine (C-C motif) ligand 8
- Gene Aliases
-
1810063B20Rik, AB023418, HC14, MCP-2, Mcp2, Scya8
- Chromosome Location
-
Chr.11: 82115185 - 82116799 on GRCm38
- UniGene ID
- Mm.42029
- Species
-
Mouse