gRNA Design Details
- Target DNA Sequence
- TGCGCATGCGATAGACTCCC
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 36779289 - 36779311 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
89.47/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Zap70
- Entrez Gene ID
-
22637
- Gene Name
- zeta-chain (TCR) associated protein kinase
- Gene Aliases
-
Srk, ZAP-70, mrtle, mur
- Chromosome Location
-
Chr.1: 36761798 - 36782821 on GRCm38
- UniGene ID
- Mm.8038
- Species
-
Mouse