gRNA Design Details
- Target DNA Sequence
- GAAAGGATACCCACACTGGC
- PAM Sequence
- CGG
- Target locus
-
Chr.3: 87906687 - 87906709 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.05/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Hdgf
- Entrez Gene ID
-
15191
- Gene Name
- hepatoma-derived growth factor
- Gene Aliases
-
AI118077, D3Ertd299e
- Chromosome Location
-
Chr.3: 87906321 - 87916132 on GRCm38
- UniGene ID
- Mm.292208
- Species
-
Mouse