gRNA Design Details
- Target DNA Sequence
- CCCATCCTTAAATAAGCTGG
- PAM Sequence
- TGG
- Target locus
-
Chr.15: 80643720 - 80643742 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
85.2/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Grap2
- Entrez Gene ID
-
17444
- Gene Name
- GRB2-related adaptor protein 2
- Gene Aliases
-
GRAP-2, GRB2L, GRBLG, GRID, Gads, GrbX, Grf40, GrpL, Mona, P38
- Chromosome Location
-
Chr.15: 80572422 - 80653047 on GRCm38
- UniGene ID
- Mm.12947
- Species
-
Mouse