gRNA Design Details
- Target DNA Sequence
- ACACTGACCAATGTCACTGG
- PAM Sequence
- GGG
- Target locus
-
Chr.18: 61061417 - 61061439 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
84.98/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Pdgfrb
- Entrez Gene ID
-
18596
- Gene Name
- platelet derived growth factor receptor, beta polypeptide
- Gene Aliases
-
AI528809, CD140b, PDGFR-1, Pdgfr
- Chromosome Location
-
Chr.18: 61045150 - 61085068 on GRCm38
- UniGene ID
- Mm.4146
- Species
-
Mouse