gRNA Design Details
- Target DNA Sequence
- GAGTGGTGATTCCCCATCCT
- PAM Sequence
- GGG
- Target locus
-
Chr.2: 28613902 - 28613924 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
85.28/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gfi1b
- Entrez Gene ID
-
14582
- Gene Name
- growth factor independent 1B
- Gene Aliases
-
Gfi-1B
- Chromosome Location
-
Chr.2: 28609450 - 28621982 on GRCm38
- UniGene ID
- Mm.373385
- Species
-
Mouse