gRNA Design Details

Target DNA Sequence
AAGATTGTGTACGACGGTCA
PAM Sequence
TGG
Target locus
Chr.13: 55156456 - 55156478 on GRCm38
Strand
Reverse
Application
Gene Knock-Out
Score: 86.19/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Fgfr4
Entrez Gene ID
14186
Gene Name
fibroblast growth factor receptor 4
Gene Aliases
Fgfr-4
Chromosome Location
Chr.13: 55152818 - 55168759 on GRCm38
UniGene ID
Mm.276715
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_008011.2 NP_032037.2 3