gRNA Design Details
- Target DNA Sequence
- TTAAGACGCGAGGCACAGGT
- PAM Sequence
- TGG
- Target locus
-
Chr.6: 122609810 - 122609832 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
92.91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gdf3
- Entrez Gene ID
-
14562
- Gene Name
- growth differentiation factor 3
- Gene Aliases
-
C78318, Gdf-3, Vgr-2, Vgr2, ecat9
- Chromosome Location
-
Chr.6: 122605403 - 122610071 on GRCm38
- UniGene ID
- Mm.299742
- Species
-
Mouse