gRNA Design Details
- Target DNA Sequence
- CGATCAAATCCCCAACAATG
- PAM Sequence
- TGG
- Target locus
-
Chr.11: 103239805 - 103239827 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
82.66/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Map3k14
- Entrez Gene ID
-
53859
- Gene Name
- mitogen-activated protein kinase kinase kinase 14
- Gene Aliases
-
Nik, aly
- Chromosome Location
-
Chr.11: 103219764 - 103267401 on GRCm38
- UniGene ID
- Mm.158981
- Species
-
Mouse