gRNA Design Details
- Target DNA Sequence
- ACGGTACTATCCAGGGAACC
- PAM Sequence
- AGG
- Target locus
-
Chr.14: 58073291 - 58073313 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
88.1/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fgf9
- Entrez Gene ID
-
14180
- Gene Name
- fibroblast growth factor 9
- Gene Aliases
-
Eks
- Chromosome Location
-
Chr.14: 58071037 - 58112720 on GRCm38
- UniGene ID
- Mm.8846
- Species
-
Mouse