gRNA Design Details
- Target DNA Sequence
- TTCGAGATTTCTTGAGGCAT
- PAM Sequence
- TGG
- Target locus
-
Chr.12: 87368976 - 87368998 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
83.06/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Sptlc2
- Entrez Gene ID
-
20773
- Gene Name
- serine palmitoyltransferase, long chain base subunit 2
- Gene Aliases
-
AI173915, LCB2, mKIAA0526
- Chromosome Location
-
Chr.12: 87307889 - 87388230 on GRCm38
- UniGene ID
- Mm.565
- Species
-
Mouse