gRNA Design Details
- Target DNA Sequence
- TCCGTGCCACAGATCAGGTT
- PAM Sequence
- GGG
- Target locus
-
Chr.4: 40929124 - 40929146 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.51/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Spink4
- Entrez Gene ID
-
20731
- Gene Name
- serine peptidase inhibitor, Kazal type 4
- Gene Aliases
-
MPGC60
- Chromosome Location
-
Chr.4: 40920056 - 40931395 on GRCm38
- UniGene ID
- Mm.25246
- Species
-
Mouse