gRNA Design Details
- Target DNA Sequence
- TCTTGTGCACTCAACAGCAG
- PAM Sequence
- GGG
- Target locus
-
Chr.4: 135344563 - 135344585 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
84.4/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Srrm1
- Entrez Gene ID
-
51796
- Gene Name
- serine/arginine repetitive matrix 1
- Gene Aliases
-
AA407769, POP101, Srm160
- Chromosome Location
-
Chr.4: 135320484 - 135353254 on GRCm38
- UniGene ID
- Mm.1963
- Species
-
Mouse