gRNA Design Details
- Target DNA Sequence
- TACCTGTGAAAAGAGACGCG
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 72824918 - 72824940 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
83.92/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Igfbp2
- Entrez Gene ID
-
16008
- Gene Name
- insulin-like growth factor binding protein 2
- Gene Aliases
-
AI255832, IBP-2, Igfbp-2, mIGFBP-2
- Chromosome Location
-
Chr.1: 72824480 - 72852474 on GRCm38
- UniGene ID
- Mm.141936
- Species
-
Mouse