gRNA Design Details
- Target DNA Sequence
- AACTCAACGCCGACGAGCTA
- PAM Sequence
- CGG
- Target locus
-
Chr.8: 70631439 - 70631461 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
92.91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gdf15
- Entrez Gene ID
-
23886
- Gene Name
- growth differentiation factor 15
- Gene Aliases
-
MIC-1, NAG-1, SBF
- Chromosome Location
-
Chr.8: 70629393 - 70632469 on GRCm38
- UniGene ID
- Mm.31325
- Species
-
Mouse