gRNA Design Details
- Target DNA Sequence
- TTTCTGACGGATCAAGCCGC
- PAM Sequence
- TGG
- Target locus
-
Chr.14: 124192483 - 124192505 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
86.18/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fgf14
- Entrez Gene ID
-
14169
- Gene Name
- fibroblast growth factor 14
- Gene Aliases
-
FHF-4, Fhf4, mFHF-4(1B)
- Chromosome Location
-
Chr.14: 123978291 - 124677666 on GRCm38
- UniGene ID
- Mm.206362
- Species
-
Mouse