gRNA Design Details
- Target DNA Sequence
- CTATGGCTACTACCAGGACG
- PAM Sequence
- AGG
- Target locus
-
Chr.11: 95578190 - 95578212 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
87.33/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ngfr
- Entrez Gene ID
-
18053
- Gene Name
- nerve growth factor receptor (TNFR superfamily, member 16)
- Gene Aliases
-
LNGFR, Tnfrsf16, p75, p75NGFR, p75NTR
- Chromosome Location
-
Chr.11: 95568818 - 95587698 on GRCm38
- UniGene ID
- Mm.283893
- Species
-
Mouse