gRNA Design Details

Target DNA Sequence
GACAGAGAACTGCCGCTCAG
PAM Sequence
TGG
Target locus
Chr.17: 12748644 - 12748666 on GRCm38
Strand
Reverse
Application
Gene Knock-Out
Score: 82.93/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Igf2r
Entrez Gene ID
16004
Gene Name
insulin-like growth factor 2 receptor
Gene Aliases
AI661837, CD222, CI-MPR, M6P/IGF2R, Mpr300
Chromosome Location
Chr.17: 12682406 - 12769706 on GRCm38
UniGene ID
Mm.26553
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_010515.2 NP_034645.2 2