gRNA Design Details
- Target DNA Sequence
- ATAGGGTACTTGTTTGCTGG
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 81905584 - 81905606 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
84.96/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Hdgfrp3
- Entrez Gene ID
-
29877
- Gene Name
- hepatoma-derived growth factor, related protein 3
- Gene Aliases
-
2700022B06Rik, HRP-3
- Chromosome Location
-
Chr.7: 81881256 - 81934459 on GRCm38
- UniGene ID
- Mm.392934
- Species
-
Mouse