gRNA Design Details
- Target DNA Sequence
- CAGAGCTCTTCAGAAACTGA
- PAM Sequence
- GGG
- Target locus
-
Chr.15: 83157539 - 83157561 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
68.1/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- HDGFRP3
- Entrez Gene ID
-
50810
- Gene Name
- hepatoma-derived growth factor, related protein 3
- Gene Aliases
-
CGI-142, HDGF-2, HDGF2, HRP-3
- Chromosome Location
-
Chr.15: 83112738 - 83208018 on GRCh38
- UniGene ID
- Hs.513954
- Species
-
Human