gRNA Design Details
- Target DNA Sequence
- GTAGCCTGATCCGACAGAAG
- PAM Sequence
- CGG
- Target locus
-
Chr.11: 69801587 - 69801609 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
93.73/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fgf11
- Entrez Gene ID
-
14166
- Gene Name
- fibroblast growth factor 11
- Gene Aliases
-
Fgf-11, Fhf-3, Fhf3
- Chromosome Location
-
Chr.11: 69796068 - 69802945 on GRCm38
- UniGene ID
- Mm.269011
- Species
-
Mouse