gRNA Design Details
- Target DNA Sequence
- AGCCTATAGGCTTTAAATTC
- PAM Sequence
- TGG
- Target locus
-
Chr.14: 33410878 - 33410900 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
83.65/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Mapk8
- Entrez Gene ID
-
26419
- Gene Name
- mitogen-activated protein kinase 8
- Gene Aliases
-
AI849689, JNK, JNK1, Prkm8, SAPK1
- Chromosome Location
-
Chr.14: 33377898 - 33447214 on GRCm38
- UniGene ID
- Mm.21495
- Species
-
Mouse