gRNA Design Details
- Target DNA Sequence
- TTGCAATCAAGAAGCTCAGC
- PAM Sequence
- CGG
- Target locus
-
Chr.14: 33408123 - 33408145 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
82.88/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Mapk8
- Entrez Gene ID
-
26419
- Gene Name
- mitogen-activated protein kinase 8
- Gene Aliases
-
AI849689, JNK, JNK1, Prkm8, SAPK1
- Chromosome Location
-
Chr.14: 33377898 - 33447214 on GRCm38
- UniGene ID
- Mm.21495
- Species
-
Mouse