gRNA Design Details
- Target DNA Sequence
- TTGGCATCAATGACATCATC
- PAM Sequence
- CGG
- Target locus
-
Chr.16: 17016029 - 17016051 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
80.27/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Mapk1
- Entrez Gene ID
-
26413
- Gene Name
- mitogen-activated protein kinase 1
- Gene Aliases
-
9030612K14Rik, AA407128, AU018647, C78273, ERK, Erk2, MAPK2, PRKM2, Prkm1, p41mapk, p42mapk
- Chromosome Location
-
Chr.16: 16983382 - 17047453 on GRCm38
- UniGene ID
- Mm.196581
- Species
-
Mouse