gRNA Design Details
- Target DNA Sequence
- GGAAAACTGCACGGTGATCG
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 68003862 - 68003884 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
87.24/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Igf1r
- Entrez Gene ID
-
16001
- Gene Name
- insulin-like growth factor I receptor
- Gene Aliases
-
A330103N21Rik, CD221, D930020L01, IGF-1R, hyft
- Chromosome Location
-
Chr.7: 67952257 - 68233668 on GRCm38
- UniGene ID
- Mm.275742
- Species
-
Mouse