gRNA Design Details
- Target DNA Sequence
- TAAATACGTGAGCTGGGTGT
- PAM Sequence
- GGG
- Target locus
-
Chr.12: 85167349 - 85167371 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.02/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Pgf
- Entrez Gene ID
-
18654
- Gene Name
- placental growth factor
- Gene Aliases
-
AI854365, PIGF, Plgf
- Chromosome Location
-
Chr.12: 85166637 - 85177785 on GRCm38
- UniGene ID
- Mm.4809
- Species
-
Mouse