gRNA Design Details
- Target DNA Sequence
- AAGCTGATCCGAGCAGGACC
- PAM Sequence
- TGG
- Target locus
-
Chr.14: 33923902 - 33923924 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
85.37/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gdf10
- Entrez Gene ID
-
14560
- Gene Name
- growth differentiation factor 10
- Gene Aliases
-
Bmp3b
- Chromosome Location
-
Chr.14: 33923587 - 33935285 on GRCm38
- UniGene ID
- Mm.432071
- Species
-
Mouse