gRNA Design Details
- Target DNA Sequence
- ACTTGCCGCAGATCTTCCTC
- PAM Sequence
- TGG
- Target locus
-
Chr.13: 56614312 - 56614334 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
84.28/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tgfbi
- Entrez Gene ID
-
21810
- Gene Name
- transforming growth factor, beta induced
- Gene Aliases
-
68kDa, AI181842, AI747162, Beta-ig, big-h3
- Chromosome Location
-
Chr.13: 56609603 - 56639339 on GRCm38
- UniGene ID
- Mm.14455
- Species
-
Mouse