gRNA Design Details
- Target DNA Sequence
- ATCGACCACGAAGTGCACGA
- PAM Sequence
- GGG
- Target locus
-
Chr.6: 55102758 - 55102780 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
92.64/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Crhr2
- Entrez Gene ID
-
12922
- Gene Name
- corticotropin releasing hormone receptor 2
- Gene Aliases
-
CRF-R2, CRFR2alpha, CRFR2beta, Crfr2
- Chromosome Location
-
Chr.6: 55090048 - 55133016 on GRCm38
- UniGene ID
- Mm.236081
- Species
-
Mouse