gRNA Design Details
- Target DNA Sequence
- AAGGAGAGCAAACCCCAAGG
- PAM Sequence
- AGG
- Target locus
-
Chr.15: 102147856 - 102147878 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
85.41/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Igfbp6
- Entrez Gene ID
-
16012
- Gene Name
- insulin-like growth factor binding protein 6
- Gene Aliases
-
IGFBP-6
- Chromosome Location
-
Chr.15: 102144186 - 102149512 on GRCm38
- UniGene ID
- Mm.358609
- Species
-
Mouse