gRNA Design Details
- Target DNA Sequence
- AGCACTAGAAGCCACGGGAG
- PAM Sequence
- TGG
- Target locus
-
Chr.7: 25687908 - 25687930 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
89.86/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tgfb1
- Entrez Gene ID
-
21803
- Gene Name
- transforming growth factor, beta 1
- Gene Aliases
-
TGF-beta1, TGFbeta1, Tgfb, Tgfb-1
- Chromosome Location
-
Chr.7: 25687002 - 25704996 on GRCm38
- UniGene ID
- Mm.248380
- Species
-
Mouse