gRNA Design Details
- Target DNA Sequence
- GTACACAGTTTCATCCTGGG
- PAM Sequence
- TGG
- Target locus
-
Chr.9: 114492948 - 114492970 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
92.15/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ccr4
- Entrez Gene ID
-
12773
- Gene Name
- chemokine (C-C motif) receptor 4
- Gene Aliases
-
C-C CKR-4, CHEMR1, Cmkbr4, LESTR, Sdf1r
- Chromosome Location
-
Chr.9: 114490316 - 114505473 on GRCm38
- UniGene ID
- Mm.1337
- Species
-
Mouse