gRNA Design Details
- Target DNA Sequence
- ACGACAAAGACCAGATCTAC
- PAM Sequence
- TGG
- Target locus
-
Chr.5: 43979592 - 43979614 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.32/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fgfbp1
- Entrez Gene ID
-
14181
- Gene Name
- fibroblast growth factor binding protein 1
- Gene Aliases
-
FGF-BP, FGF-BP1, FGFBP-1
- Chromosome Location
-
Chr.5: 43978858 - 43981827 on GRCm38
- UniGene ID
- Mm.46053
- Species
-
Mouse