gRNA Design Details

Target DNA Sequence
CGCCGTGATCAGTTGGACTA
PAM Sequence
AGG
Target locus
Chr.10: 121565591 - 121565613 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 76.39/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
FGFR2
Entrez Gene ID
2263
Gene Name
fibroblast growth factor receptor 2
Gene Aliases
BBDS, BEK, BFR-1, CD332, CEK3, CFD1, ECT1, JWS, K-SAM, KGFR, TK14, TK25
Chromosome Location
Chr.10: 121478330 - 121598656 on GRCh38
UniGene ID
Hs.533683
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000141.4 NP_000132.3 3
NM_001144913.1 NP_001138385.1 2
NM_001144914.1 NP_001138386.1 2
NM_001144917.1 NP_001138389.1 3
NM_001320658.1 NP_001307587.1 3
NM_022970.3 NP_075259.4 3
XM_006717708.2 XP_006717771.1 3
XM_006717710.3 XP_006717773.1 3
XM_017015920.1 XP_016871409.1 3
XM_017015921.1 XP_016871410.1 3