gRNA Design Details
- Target DNA Sequence
- GTGCCCTTAGTACCAAGCTC
- PAM Sequence
- GGG
- Target locus
-
Chr.15: 86033434 - 86033456 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
85.88/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Celsr1
- Entrez Gene ID
-
12614
- Gene Name
- cadherin, EGF LAG seven-pass G-type receptor 1 (flamingo homolog, Drosophila)
- Gene Aliases
-
Adgrc1, Crsh, Scy, crash
- Chromosome Location
-
Chr.15: 85898758 - 86034217 on GRCm38
- UniGene ID
- Mm.22680
- Species
-
Mouse