gRNA Design Details
- Target DNA Sequence
- TCGGAACAGACTGTACACCT
- PAM Sequence
- TGG
- Target locus
-
Chr.12: 84876013 - 84876035 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
85/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ltbp2
- Entrez Gene ID
-
16997
- Gene Name
- latent transforming growth factor beta binding protein 2
- Gene Aliases
-
AW208642
- Chromosome Location
-
Chr.12: 84783212 - 84877266 on GRCm38
- UniGene ID
- Mm.3900
- Species
-
Mouse