gRNA Design Details
- Target DNA Sequence
- TCTATACATGGAGACACACC
- PAM Sequence
- TGG
- Target locus
-
Chr.8: 70498662 - 70498684 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.7/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Crlf1
- Entrez Gene ID
-
12931
- Gene Name
- cytokine receptor-like factor 1
- Gene Aliases
-
CLF-1, CRLM3, NR6.1
- Chromosome Location
-
Chr.8: 70493107 - 70504081 on GRCm38
- UniGene ID
- Mm.335584
- Species
-
Mouse