gRNA Design Details
- Target DNA Sequence
- AGTGATGTTCATCTTCGGGG
- PAM Sequence
- TGG
- Target locus
-
Chr.15: 5243040 - 5243062 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.16/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ptger4
- Entrez Gene ID
-
19219
- Gene Name
- prostaglandin E receptor 4 (subtype EP4)
- Gene Aliases
-
EP4, Ptgerep4
- Chromosome Location
-
Chr.15: 5233399 - 5244187 on GRCm38
- UniGene ID
- Mm.18509
- Species
-
Mouse