gRNA Design Details
- Target DNA Sequence
- GATGTGGCCGAAGTCCAACG
- PAM Sequence
- TGG
- Target locus
-
Chr.12: 86077828 - 86077850 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
92.64/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tgfb3
- Entrez Gene ID
-
21809
- Gene Name
- transforming growth factor, beta 3
- Gene Aliases
-
Tgfb-3
- Chromosome Location
-
Chr.12: 86056743 - 86079041 on GRCm38
- UniGene ID
- Mm.3992
- Species
-
Mouse