gRNA Design Details

Target DNA Sequence
TGATTGTGGCCAAGTTCAGC
PAM Sequence
AGG
Target locus
Chr.12: 86077872 - 86077894 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 88.36/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Tgfb3
Entrez Gene ID
21809
Gene Name
transforming growth factor, beta 3
Gene Aliases
Tgfb-3
Chromosome Location
Chr.12: 86056743 - 86079041 on GRCm38
UniGene ID
Mm.3992
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_009368.3 NP_033394.2 1