gRNA Design Details
- Target DNA Sequence
- GCTCAGTAGCTCTGACTGTG
- PAM Sequence
- GGG
- Target locus
-
Chr.7: 45421694 - 45421716 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
82.68/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Lhb
- Entrez Gene ID
-
16866
- Gene Name
- luteinizing hormone beta
- Gene Aliases
-
LH, LH-B, LSH-B, LSH-beta
- Chromosome Location
-
Chr.7: 45418166 - 45421855 on GRCm38
- UniGene ID
- Mm.57061
- Species
-
Mouse