gRNA Design Details
- Target DNA Sequence
- CTCAGTCTGCACCAATATCC
- PAM Sequence
- TGG
- Target locus
-
Chr.17: 57402544 - 57402566 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
83.45/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Adgre1
- Entrez Gene ID
-
13733
- Gene Name
- adhesion G protein-coupled receptor E1
- Gene Aliases
-
DD7A5-7, EGF-TM7, Emr1, F4/80, Gpf480, Ly71, TM7LN3
- Chromosome Location
-
Chr.17: 57358651 - 57483529 on GRCm38
- UniGene ID
- Mm.2254
- Species
-
Mouse