gRNA Design Details
- Target DNA Sequence
- GGACTAGAGCAAGCAATGAC
- PAM Sequence
- AGG
- Target locus
-
Chr.11: 83525917 - 83525939 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
85.72/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ccl5
- Entrez Gene ID
-
20304
- Gene Name
- chemokine (C-C motif) ligand 5
- Gene Aliases
-
MuRantes, RANTES, SISd, Scya5, TCP228
- Chromosome Location
-
Chr.11: 83525778 - 83530518 on GRCm38
- UniGene ID
- Mm.284248
- Species
-
Mouse